Tris(3-aminopropyl)amine ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
N1,N1-Bis(3-aminopropyl)propane-1,3-diamine, 3,3',3''-Nitrilotris(propylamine)
T1540-25ML
T1540-5ML
TCIAT1540-25MLEA
697
EUR
TCIAT1540-25ML
TCIAT1540-5ML
Tris(3-aminopropyl)amine ≥97.0% (by titrimetric analysis)
Tris(3-aminopropyl)amine
Formule:
C₉H₂₄N₄ MW: 188.32 g/mol Kookpunt: 150°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00191593 CAS nummer: 4963-47-7 UN: 2735 ADR: 8 III |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...