Tetraethyl-p-xylylenediphosphonate ≥95.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: p-Xylylenediphosphonic Acid Tetraethyl Ester, p-Bis(diethylphosphono)xylene

T1582-100G T1582-25G T1582-5G
TCIAT1582-100GEA 247 EUR
TCIAT1582-100G TCIAT1582-25G TCIAT1582-5G
Tetraethyl-p-xylylenediphosphonate ≥95.0% (by HPLC)
Tetraethyl-p-xylylenediphosphonate
Formule: C₁₆H₂₈O₆P₂
MW: 378.34 g/mol
Kookpunt: 220°C
MDL Nummer: MFCD00671576
CAS nummer: 4546-04-7
EINECS: 224-902-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 73.0 to 78.0 deg-C
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR