Tetrabutylammonium difluorotriphenylstannate ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T1592-1G T1592-5G
TCIAT1592-1GEA 52.8 EUR
TCIAT1592-1G TCIAT1592-5G
Tetrabutylammonium difluorotriphenylstannate ≥97.0% (by titrimetric analysis)
Tetrabutylammonium difluorotriphenylstannate
Formule: C₃₄H₅₁F₂NSn
MW: 630.50 g/mol
MDL Nummer: MFCD00192465
CAS nummer: 139353-88-1
UN: 3146
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 191.0 to 195.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR