Dimethyl tetrahydropyran-4,4-dicarboxylate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Tetrahydropyran-4,4-dicarboxylic Acid Dimethyl Ester, Dimethyl Tetrahydropyran-4,4-dicarboxylate

T1825-25G T1825-5G
TCIAT1825-25GEA 411 EUR
TCIAT1825-25G TCIAT1825-5G
Dimethyl tetrahydropyran-4,4-dicarboxylate ≥98.0% (by GC)
Dimethyl tetrahydropyran-4,4-dicarboxylate
Formule: C₉H₁₄O₅
MDL Nummer: MFCD02093499
CAS nummer: 149777-00-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR