Tetrabutylammonium difluorotriphenylsilicate ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: TBAT, Tetrabutylammonium difluorotriphenylsilicate(IV)

T1909-25G T1909-5G
TCIAT1909-25GEA 293 EUR
TCIAT1909-25G TCIAT1909-5G
Tetrabutylammonium difluorotriphenylsilicate ≥97.0% (by titrimetric analysis)
Tetrabutylammonium difluorotriphenylsilicate
Formule: C₃₄H₅₁F₂NSi
MW: 539.87 g/mol
MDL Nummer: MFCD00274218
CAS nummer: 163931-61-1
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR