Tri-o-tolylbismuthine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

T1970-5G T1970-1G
TCIAT1970-5GEA 131 EUR
TCIAT1970-5G TCIAT1970-1G
Tri-o-tolylbismuthine ≥98.0% (by HPLC, titration analysis)
Tri-o-tolylbismuthine
Formule: C₂₁H₂₁Bi
MDL Nummer: MFCD00123270
CAS nummer: 10050-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efadd8fa2bdbee608206a83ca99b842bd342ca4e&vid=efadd8fa2bdbee608206a83ca99b842bd342ca4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Chelometric Titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C
Solubility in THF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR