1,5,7-Triazabicyclo[4.4.0]dec-5-ene ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, Hhpp, 2,3,4,6,7,8-Hexahydro-1H-pyrimido[1,2-a]pyrimidine, TBD

T1982-5G T1982-25G
TCIAT1982-5GEA 84.7 EUR
TCIAT1982-5G TCIAT1982-25G
1,5,7-Triazabicyclo[4.4.0]dec-5-ene ≥98.0% (by HPLC, titration analysis)
1,5,7-Triazabicyclo[4.4.0]dec-5-ene
Formule: C₇H₁₃N₃
MW: 139.20 g/mol
MDL Nummer: MFCD00043003
CAS nummer: 5807-14-7
EINECS: 227-367-1
UN: 3263
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92de1097668a32c7d3a33d818e95d77662419597&vid=92de1097668a32c7d3a33d818e95d77662419597&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR