2,4,5,6-Tetrafluoro-1,3-phenylenediamine ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1,3-Diamino-2,4,5,6-tetrafluorobenzene

T2019-1G T2019-5G
TCIAT2019-1GEA 104 EUR
TCIAT2019-1G TCIAT2019-5G
2,4,5,6-Tetrafluoro-1,3-phenylenediamine ≥95.0% (by GC)
2,4,5,6-Tetrafluoro-1,3-phenylenediamine
Formule: C₆H₄F₄N₂
Smeltpunt: 131°C
MDL Nummer: MFCD00039764
CAS nummer: 1198-63-6
EINECS: 214-836-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR