(3-Isocyanatopropyl)trimethoxysilane ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T2044-100G T2044-25G
TCIAT2044-100GEA 108 EUR
TCIAT2044-100G TCIAT2044-25G
(3-Isocyanatopropyl)trimethoxysilane ≥97.0% (by GC)
(3-Isocyanatopropyl)trimethoxysilane
Formule: C₇H₁₅NO₄Si
Kookpunt: 75°C
Vlampunt: 108°C
MDL Nummer: MFCD00015793
CAS nummer: 15396-00-6
EINECS: 239-415-9
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR