Titanyl Phthalocyanine, purified by sublimation

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T2272-200MG T2272-1G
TCIAT2272-200MGEA 61.6 EUR
TCIAT2272-200MG TCIAT2272-1G
Titanyl Phthalocyanine, purified by sublimation
Titanyl Phthalocyanine
Formule: C₃₂H₁₆N₈OTi
Smeltpunt: 395°C
MDL Nummer: MFCD00145414
CAS nummer: 26201-32-1
EINECS: 419-970-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=657e602e26a8ca5d08cd58d5774bba735de9389e&vid=657e602e26a8ca5d08cd58d5774bba735de9389e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Dark red to Dark purple to Dark blue powder to crystal
Elemental analysis (Nitrogen) 18.50 to 20.00 %
Lambda max. 692.0 to 694.0 nm (Pyridine)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR