3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 3,3,5-Trimethylcyclohexyl 2-hydroxybenzoate, Coppertone, 2-Hydroxybenzoic Acid 3,3,5-Trimethylcyclohexyl Ester, Salicylic Acid 3,3,5-Trimethylcyclohexyl Ester, 3,3,5-Trimethylcyclohexyl 2-Hydroxybenzoate, Homosalate

T2278-25G T2278-500G T2278-100G
TCIAT2278-25GEA 17.6 EUR
TCIAT2278-25G TCIAT2278-500G TCIAT2278-100G
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture)
Formule: C₁₆H₂₂O₃
Kookpunt: 165°C
MDL Nummer: MFCD00019377
CAS nummer: 118-56-9
EINECS: 204-260-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=434d2a50f3a0a6c75f78e0a6037aeefb9ec75ae7&vid=434d2a50f3a0a6c75f78e0a6037aeefb9ec75ae7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR