2,3-Thiophenedicarboxaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T2388-1G
T2388-5G
TCIAT2388-1GEA
91.3
EUR
TCIAT2388-1G
TCIAT2388-5G
2,3-Thiophenedicarboxaldehyde ≥98.0% (by GC)
2,3-Thiophenedicarboxaldehyde
Formule:
C₆H₄O₂S MW: 140.16 g/mol Kookpunt: 134°C |
MDL Nummer:
MFCD00209616 CAS nummer: 932-41-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...