1,1,3-Triphenyl-2-propyn-1-ol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T2418-25G T2418-5G
TCIAT2418-25GEA 140 EUR
TCIAT2418-25G TCIAT2418-5G
1,1,3-Triphenyl-2-propyn-1-ol ≥98.0% (by GC)
1,1,3-Triphenyl-2-propyn-1-ol
Formule: C₂₁H₁₆O
Kookpunt: 220°C
MDL Nummer: MFCD00004449
CAS nummer: 1522-13-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR