D-Tryptophan benzyl ester hydrochloride ≥98.0% (by total nitrogen basis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
H-D-Trp-OBzl·HCl, H-D-Trp-OBzl.HCl
T2438-1G
T2438-5G
TCIAT2438-1GEA
68.2
EUR
TCIAT2438-1G
TCIAT2438-5G
D-Tryptophan benzyl ester hydrochloride ≥98.0% (by total nitrogen basis)
D-Tryptophan benzyl ester hydrochloride
Formule:
C₁₈H₁₈N₂O₂ Smeltpunt: 215°C |
MDL Nummer:
MFCD00237293 CAS nummer: 22839-16-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | -30.0 to -35.0 deg (C=1, DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...