4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T2471-1G
T2471-5G
TCIAT2471-1GEA
89.1
EUR
TCIAT2471-1G
TCIAT2471-5G
4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one ≥98.0% (by GC)
4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one
Formule:
C₈H₈O₄ |
CAS nummer:
632339-23-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 179.0 to 183.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...