1,1,3,3-Tetrabutylurea ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

T2537-25G T2537-5G
TCIAT2537-25GEA 113 EUR
TCIAT2537-25G TCIAT2537-5G
1,1,3,3-Tetrabutylurea ≥98.0% (by GC)
1,1,3,3-Tetrabutylurea
Formule: C₁₇H₃₆N₂O
MW: 284.49 g/mol
Kookpunt: 163°C
Vlampunt: 93°C
MDL Nummer: MFCD00042901
CAS nummer: 4559-86-8
EINECS: 224-929-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR