Trimethyl(vinyloxy)silane ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T2561-1G T2561-5G
TCIAT2561-1GEA 25.3 EUR
TCIAT2561-1G TCIAT2561-5G
Trimethyl(vinyloxy)silane ≥95.0% (by GC)
Trimethyl(vinyloxy)silane
Formule: C₅H₁₂OSi
Kookpunt: 75°C
Vlampunt: -19°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00054764
CAS nummer: 6213-94-1
EINECS: 228-281-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR