4,4,5,5-Tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T2572-25G T2572-5G
TCIAT2572-25GEA 149 EUR
TCIAT2572-25G TCIAT2572-5G
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC)
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Formule: C₆H₁₃BO₂
Kookpunt: 43°C
Vlampunt: 5°C
MDL Nummer: MFCD00674030
CAS nummer: 25015-63-8
UN: 3399
ADR: 4.3,II
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %