Trimethylsilyl trifluoroacetate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Trifluoroacetic Acid Trimethylsilyl Ester
T2629-25G T2629-5G
TCIAT2629-25GEA 156 EUR
TCIAT2629-25G TCIAT2629-5G
Trimethylsilyl trifluoroacetate ≥95.0% (by GC)
Trimethylsilyl trifluoroacetate
Formule: C₅H₉F₃O₂Si
MW: 186.21 g/mol
Kookpunt: 90°C
Vlampunt: 13°C
MDL Nummer: MFCD00000413
CAS nummer: 400-53-3
EINECS: 206-923-7
UN: 2924
ADR: 3 II
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %