Topiramate ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2,3:4,5-Di-O-isopropylidene-1-O-sulphamoyl-beta-D-fructopyranose, ((3aS,5aR,8aR,8bS)-2,2,7,7-Tetramethyltetrahydro-3aH-bis([1,3]dioxolo)[4,5-b:4',5'-d]pyran-3a-yl)methyl sulphamate
T2755-1G
T2755-5G
TCIAT2755-1GEA
45.1
EUR
TCIAT2755-1G
TCIAT2755-5G
Topiramate ≥98.0% (by HPLC, titration analysis)
Topiramate
Formule:
C₁₂H₂₁NO₈S Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00865320 CAS nummer: 97240-79-4 Merck Index: 14,09547 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 124.0 to 128.0 deg-C |
Specific rotation [a]20/D | -29.0 to -33.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...