α-Chloro-2,4,5-trifluorotoulene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 2,4,5-Trifluorobenzyl chloride, 1-(Chloromethyl)-2,4,5-trifluorobenzene

T2889-1G T2889-5G
TCIAT2889-1GEA 37.4 EUR
TCIAT2889-1G TCIAT2889-5G
α-Chloro-2,4,5-trifluorotoulene ≥98.0% (by GC)
α-Chloro-2,4,5-trifluorotoulene
Formule: C₇H₄ClF₃
Vlampunt: 65°C
MDL Nummer: MFCD00236319
CAS nummer: 243139-71-1
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=717c75157756ccb776516fcb9aaf725bfb977c39&vid=717c75157756ccb776516fcb9aaf725bfb977c39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR