Trimethylamine ∼25% ∼3 mol/L in isopropanol (IPA)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T2893-100ML T2893-500ML
TCIAT2893-100MLEA 23.1 EUR
TCIAT2893-100ML TCIAT2893-500ML
Trimethylamine ∼25% ∼3 mol/L in isopropanol (IPA)
Trimethylamine
Formule: C₃H₉N
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008327
CAS nummer: 75-50-3
EINECS: 200-875-0
UN: 2924
ADR: 3,II
Merck Index: 14,09710
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2c47a70c7f2f26751c1c18b1d7821251f630f2b&vid=c9498b23da3a46e02da4058ff2a2df8463f17fd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (Neutralization titration) 24.5 to 27.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR