Triisopropanolamine cyclic borate ≥98.0% (by total nitrogen basis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T2943-5G
T2943-25G
TCIAT2943-5GEA
79.2
EUR
TCIAT2943-5G
TCIAT2943-25G
Triisopropanolamine cyclic borate ≥98.0% (by total nitrogen basis)
Triisopropanolamine cyclic borate
Formule:
C₉H₁₈BNO₃ Kookpunt: 135°C |
MDL Nummer:
MFCD03791298 CAS nummer: 101-00-8 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...