Tetrahydroxydiborane
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T2953-5G
T2953-25G
TCIAT2953-5GEA
78.1
EUR
TCIAT2953-5G
TCIAT2953-25G
Tetrahydroxydiborane
Tetrahydroxydiborane
Formule:
H₄B₂O₄ Smeltpunt: 290°C |
MDL Nummer:
MFCD05663888 CAS nummer: 13675-18-8 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 98.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...