Tetrahydropyran-4-methanol ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-(Hydroxymethyl)tetrahydropyran
T2965-1G T2965-5G
TCIAT2965-1GEA 39.6 EUR
TCIAT2965-1G TCIAT2965-5G
Tetrahydropyran-4-methanol ≥97.0% (by GC)
Tetrahydropyran-4-methanol
Formule: C₆H₁₂O₂
MW: 116.16 g/mol
Kookpunt: 110°C
MDL Nummer: MFCD00457804
CAS nummer: 14774-37-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b64d6cf3e40ace8819d5ad252096daa6762b1132&vid=b64d6cf3e40ace8819d5ad252096daa6762b1132&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %