1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Allylhexafluoroisopropanol
T2968-5G T2968-25G
TCIAT2968-5GEA 68.2 EUR
TCIAT2968-5G TCIAT2968-25G
1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol ≥98.0% (by GC)
1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol
Formule: C₆H₆F₆O
Kookpunt: 100°C
MDL Nummer: MFCD03092986
CAS nummer: 646-97-9
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %