(Triphenylphosphine)gold(I) chloride ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold

T2994-200MG T2994-1G
TCIAT2994-200MGEA 63.8 EUR
TCIAT2994-200MG TCIAT2994-1G
(Triphenylphosphine)gold(I) chloride ≥95.0% (by titrimetric analysis)
(Triphenylphosphine)gold(I) chloride
Formule: C₁₈H₁₅AuClP
MW: 494.71 g/mol
MDL Nummer: MFCD00009588
CAS nummer: 14243-64-2
EINECS: 238-117-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92d00d2713b290555cbd46787438f58b10495212&vid=92d00d2713b290555cbd46787438f58b10495212&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 95.0 %
Melting point 245.0 to 249.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR