1,2,3,4-Tetrahydroquinoxaline ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T2999-1G T2999-5G
TCIAT2999-1GEA 68.2 EUR
TCIAT2999-1G TCIAT2999-5G
1,2,3,4-Tetrahydroquinoxaline ≥98.0% (by GC, titration analysis)
1,2,3,4-Tetrahydroquinoxaline
Formule: C₈H₁₀N₂
Kookpunt: 154°C
MDL Nummer: MFCD00047564
CAS nummer: 3476-89-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26bb13c19dba29334db9a1e45b7989aba7013a8d&vid=26bb13c19dba29334db9a1e45b7989aba7013a8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR