Tenofovir ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (R)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid, (R)-[[2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid

T3006-1G T3006-5G
TCIAT3006-1GEA 17.6 EUR
TCIAT3006-1G TCIAT3006-5G
Tenofovir ≥98.0% (by HPLC, titration analysis)
Tenofovir
Formule: C₉H₁₄N₅O₄P
MW: 287.22 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00943794
CAS nummer: 147127-20-6
Merck Index: 14,09146
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd128d963daa7df2acffdba83efa5c0c396231ff&vid=bd128d963daa7df2acffdba83efa5c0c396231ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -19.0 to -21.0 deg (C=1, 0.1mol/L HCl) (calcd.on anh.substance)
Water max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR