4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T3027-5G T3027-25G
TCIAT3027-5GEA 61.6 EUR
TCIAT3027-5G TCIAT3027-25G
4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid ≥98.0% (by GC, titration analysis)
4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Formule: C₉H₆F₄O₃
MDL Nummer: MFCD00155926
CAS nummer: 10009-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecd461009b44069110178270846baa42eaa68464&vid=ecd461009b44069110178270846baa42eaa68464&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 176.0 to 180.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR