Tris(phenylthio)methane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T3126-5G T3126-25G
TCIAT3126-5GEA 55 EUR
TCIAT3126-5G TCIAT3126-25G
Tris(phenylthio)methane ≥98.0% (by GC)
Tris(phenylthio)methane
Formule: C₁₉H₁₆S₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00010396
CAS nummer: 4832-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=794f6f4ba9f42171996d0880d2b3b48e7686be7a&vid=794f6f4ba9f42171996d0880d2b3b48e7686be7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR