Tianeptine sodium salt hydrate ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
7-[[3-Chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-yl]amino]heptanoic acid S,S-dioxide sodium salt hydrate
T3131-200MG
T3131-1G
TCIAT3131-200MGEA
78.1
EUR
TCIAT3131-200MG
TCIAT3131-1G
Tianeptine sodium salt hydrate ≥98.0% (by HPLC, total nitrogen)
Tianeptine sodium salt hydrate
Formule:
C₂₁H₂₄ClN₂NaO₄S·xH₂O MW: 458.93 g/mol (anhy) |
MDL Nummer:
MFCD00879851 CAS nummer: 30123-17-2 EINECS: 250-059-3 Merck Index: 14,09420 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % (calcd.on anh.substance) |
Melting point | 183.0 to 187.0 deg-C |
Water | 3.0 to 7.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...