1-(Trifluoromethanesulphonyl)imidazole ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-((Trifluoromethyl)sulphonyl)-1H-imidazole
T3177-1G T3177-5G
TCIAT3177-1GEA 154 EUR
TCIAT3177-1G TCIAT3177-5G
1-(Trifluoromethanesulphonyl)imidazole ≥98.0% (by GC)
1-(Trifluoromethanesulphonyl)imidazole
Formule: C₄H₃F₃N₂O₂S
MW: 200.14 g/mol
Kookpunt: 40°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00037891
CAS nummer: 29540-81-6
EINECS: 249-686-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4239625f996bee3e4e2bbbf40344f12332131261&vid=4239625f996bee3e4e2bbbf40344f12332131261&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %