1H,1H,2H,2H-Perfluorooctyl methacrylate ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Methacrylic acid 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-n-octyl ester, 2-(Perfluorohexyl)ethyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-n-octyl Methacrylate, 1H,1H,2H,2H-Tridecafluoro-n-octyl Methacrylate, Methacrylic acid 1H,1H,2H,2H-Tridecafluoro-n-octyl ester
Stabilisatie:
Stabilised with HQ + MEHQ
T3414-5G
T3414-25G
T3414-100G
TCIAT3414-5GEA
50.6
EUR
TCIAT3414-5G
TCIAT3414-25G
TCIAT3414-100G
1H,1H,2H,2H-Perfluorooctyl methacrylate ≥98.0% (by GC) stabilised
1H,1H,2H,2H-Perfluorooctyl methacrylate
Formule:
C₁₂H₉F₁₃O₂ MW: 432.18 g/mol |
MDL Nummer:
MFCD00077580 CAS nummer: 2144-53-8 EINECS: 218-407-9 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...