Tris(2-chloorethyl)fosfaat 93,0 GC_ASSAY_METHOD

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Phosphoric acid tris(2-chloroethyl) ester

T3447-25G T3447-100G
TCIAT3447-25GEA 27.5 EUR
TCIAT3447-25G TCIAT3447-100G
Tris(2-chloorethyl)fosfaat 93,0 GC_ASSAY_METHOD
Tris(2-chloorethyl)fosfaat
Formule: C₆H₁₂Cl₃O₄P
MW: 285.49 g/mol
Kookpunt: 146°C
Vlampunt: 222°C
MDL Nummer: MFCD00000967
CAS nummer: 115-96-8
EINECS: 204-118-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25088439389faa1275d946e1da6b35c011d96375&vid=25088439389faa1275d946e1da6b35c011d96375&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR