4-(Morpholinomethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester, 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 4,4,5,5-Tetramethyl-2-[4-(morpholinomethyl)phenyl]-1,3,2-dioxaborolane, 4-(Morpholinomethyl)phenylboronic acid Pinacol ester
T3477-1G
T3477-5G
TCIAT3477-1GEA
37.4
EUR
TCIAT3477-1G
TCIAT3477-5G
4-(Morpholinomethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Morpholinomethyl)phenylboronic acid pinacol ester
Formule:
C₁₇H₂₆BNO₃ |
MDL Nummer:
MFCD04974052 CAS nummer: 364794-79-6 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...