4-(Trifluoromethyl)cyclohexylamine cis- and trans-mixture ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T3613-1G
T3613-5G
TCIAT3613-1GEA
69.3
EUR
TCIAT3613-1G
TCIAT3613-5G
4-(Trifluoromethyl)cyclohexylamine cis- and trans-mixture ≥98.0% (by GC)
4-(Trifluoromethyl)cyclohexylamine
Formule:
C₇H₁₂F₃N Kookpunt: 71°C |
MDL Nummer:
MFCD09040573 CAS nummer: 58665-70-6 UN: 2734 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...