Gallium(III) acetylacetonate ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T3652-5G T3652-25G
TCIAT3652-5GEA 69.3 EUR
TCIAT3652-5G TCIAT3652-25G
Gallium(III) acetylacetonate ≥97.0% (by titrimetric analysis)
Gallium(III) acetylacetonate
Formule: C₁₅H₂₁GaO₆
Smeltpunt: 200°C
MDL Nummer: MFCD00013492
CAS nummer: 14405-43-7
EINECS: 238-377-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7cc9cf3433f8c6d67014947a0374a2b662a8ad0&vid=c7cc9cf3433f8c6d67014947a0374a2b662a8ad0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystaline
Purity (Chelometric Titration) min. 97.0 %
Melting point 196.0 to 200.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR