Tetraisocyanatosilane ≥97.0% (by total nitrogen basis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T3684-5G
T3684-25G
TCIAT3684-5GEA
74.8
EUR
TCIAT3684-5G
TCIAT3684-25G
Tetraisocyanatosilane ≥97.0% (by total nitrogen basis)
Tetraisocyanatosilane
Formule:
C₄N₄O₄Si Kookpunt: 186°C Smeltpunt: 26°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00040537 CAS nummer: 3410-77-3 EINECS: 222-297-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...