Tetradecamethylhexasiloxane ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
1,1,1,3,3,5,5,7,7,9,9,11,11,11-Tetradecamethylhexasiloxane
T3685-5ML
T3685-25ML
TCIAT3685-5MLEA
62.7
EUR
TCIAT3685-5ML
TCIAT3685-25ML
Tetradecamethylhexasiloxane ≥96.0% (by GC)
Tetradecamethylhexasiloxane
Formule:
C₁₄H₄₂O₅Si₆ MW: 459.00 g/mol Kookpunt: 142°C Smeltpunt: –59°C Vlampunt: 118°C |
MDL Nummer:
MFCD08275354 CAS nummer: 107-52-8 EINECS: 203-499-5 Merck Index: 14,09197 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...