Tris(4-bromophenyl)amine ≥99.0% (by GC), purified by sublimation

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T3777-200MG T3777-1G
TCIAT3777-200MGEA 53.9 EUR
TCIAT3777-200MG TCIAT3777-1G
Tris(4-bromophenyl)amine ≥99.0% (by GC), purified by sublimation
Tris(4-bromophenyl)amine
Formule: C₁₈H₁₂Br₃N
MW: 482.01 g/mol
MDL Nummer: MFCD00009665
CAS nummer: 4316-58-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 144.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR