Tissue-clearing reagent, CUBIC-HL for animals
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T3781-100ML T3781-25ML
TCIAT3781-100MLEA 143 EUR
TCIAT3781-100ML TCIAT3781-25ML
Tissue-clearing reagent, CUBIC-HL for animals
Kleurstoffen Microscopy and staining reagents
Specifications
Appearance: Colorless to Light yellow to Light orange clear liquid
Decoloring test: min. 1.0 (400 to 410 nm)