Tissue-clearing reagent, CUBIC-P for animals
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T3782-100ML
T3782-25ML
TCIAT3782-100MLEA
143
EUR
TCIAT3782-100ML
TCIAT3782-25ML
Tissue-clearing reagent, CUBIC-P for animals
Kleurstoffen
Microscopy and staining reagents
Specifications
Appearance: Colorless to Almost colorless clear liquid
Functionality test: to pass test (Remaining fluorescence strength)
Decoloring test: min. 1.0 (400 to 410 nm)