Triisopropylsilyl acrylate ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

T3789-25G T3789-5G
TCIAT3789-25GEA 143 EUR
TCIAT3789-25G TCIAT3789-5G
Triisopropylsilyl acrylate ≥98.0% (by GC) stabilised
Triisopropylsilyl acrylate
Formule: C₁₂H₂₄O₂Si
Kookpunt: 102°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01861733
CAS nummer: 157859-20-6
EINECS: 457-670-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR