2-([1,1':3',1''-Terphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T3828-1G
T3828-250MG
TCIAT3828-1GEA
200
EUR
TCIAT3828-1G
TCIAT3828-250MG
2-([1,1':3',1''-Terphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-([1,1':3',1''-Terphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formule:
C₂₄H₂₅BO₂ |
MDL Nummer:
MFCD28134531 CAS nummer: 1115023-84-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...