2,2,5-Trimethyl-1,3-dioxane-5-carboxylic acid ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T3839-5G T3839-25G
TCIAT3839-5GEA 101 EUR
TCIAT3839-5G TCIAT3839-25G
2,2,5-Trimethyl-1,3-dioxane-5-carboxylic acid ≥95.0% (by titrimetric analysis)
2,2,5-Trimethyl-1,3-dioxane-5-carboxylic acid
Formule: C₈H₁₄O₄
MDL Nummer: MFCD08272187
CAS nummer: 16837-14-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR