Tissue-clearing reagent, CUBIC-X2 for animals
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T3867-25ML
T3867-100ML
TCIAT3867-25MLEA
45.1
EUR
TCIAT3867-25ML
TCIAT3867-100ML
Tissue-clearing reagent, CUBIC-X2 for animals
Kleurstoffen
Microscopy and staining reagents
Specifications
Appearance: Colorless to Light yellow to Light orange clear liquid
Functionality test: to pass test (Remaining fluorescence strength)