N-Succinimidyl N-methylcarbamate ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-Succinimidyl methyllcarbamate, Methylcarbamic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl methylcarbamate

U0108-1G U0108-5G
TCIAU0108-1GEA 29.7 EUR
TCIAU0108-1G TCIAU0108-5G
N-Succinimidyl N-methylcarbamate ≥98.0% (by total nitrogen basis)
N-Succinimidyl N-methylcarbamate
Formule: C₆H₈N₂O₄
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00800292
CAS nummer: 18342-66-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR