5-Vinylbicyclo[2.2.1]hept-2-ene ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Stabilisatie:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
V0062-25ML
V0062-500ML
TCIAV0062-25MLEA
17.6
EUR
TCIAV0062-25ML
TCIAV0062-500ML
5-Vinylbicyclo[2.2.1]hept-2-ene ≥98.0% (by GC) stabilised
5-Vinylbicyclo[2.2.1]hept-2-ene
Formule:
C₉H₁₂ Kookpunt: 141°C Vlampunt: 22°C |
MDL Nummer:
MFCD00080691 CAS nummer: 3048-64-4 EINECS: 221-259-8 UN: 1992 ADR: 3,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...