L-Valinamide hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

V0102-5G V0102-25G
TCIAV0102-5GEA 39.6 EUR
TCIAV0102-5G TCIAV0102-25G
L-Valinamide hydrochloride ≥98.0% (by total nitrogen and titration analysis)
L-Valinamide hydrochloride
Formule: C₅H₁₂N₂O
Smeltpunt: 263°C
MDL Nummer: MFCD00039085
CAS nummer: 3014-80-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83bf911fedd98c60406cf36b2d710fa2da968cf0&vid=83bf911fedd98c60406cf36b2d710fa2da968cf0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +24.0 to +30.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR